Assay Details
| Gene Symbol: OST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005744.3 | XP_004005793.2 | 1 - 2 | 116 | 79 | |
| XM_012166314.2 | XP_012021704.1 | 1 - 2 | 116 | 79 | ||
More Information
Set Membership: |
|

OST4
Genomic Map