Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178747.2 | XP_012034137.1 | 5 - 6 | 573 | 58 | |
| XM_027973230.1 | XP_027829031.1 | 5 - 6 | 573 | 58 | ||
More Information
Set Membership: |
|

TAFA3
Genomic Map