Assay Details
| Gene Symbol: AKAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972558.1 | XP_027828359.1 | 4 - 5 | 5481 | 64 | |
| XM_027972560.1 | XP_027828361.1 | 2 - 3 | 5461 | 64 | ||
More Information
Set Membership: |
|

AKAP12
Genomic Map