Assay Details
| Gene Symbol: LOC101104843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975767.1 | XP_027831568.1 | 11 - 12 | 1762 | 73 | |
| XM_027975770.1 | XP_027831571.1 | 11 - 12 | 1764 | 73 | ||
| XM_027975771.1 | XP_027831572.1 | 11 - 12 | 1763 | 73 | ||
| XM_027975772.1 | XP_027831573.1 | 10 - 11 | 1565 | 73 | ||
| XM_027975773.1 | XP_027831574.1 | 2 - 3 | 212 | 73 | ||
More Information
Set Membership: |
|

LOC101104843
Genomic Map