Assay Details
| Gene Symbol: LOC101104843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975767.1 | XP_027831568.1 | 25 - 26 | 3446 | 76 | |
| XM_027975770.1 | XP_027831571.1 | 25 - 26 | 3445 | 76 | ||
| XM_027975771.1 | XP_027831572.1 | 25 - 26 | 3417 | 76 | ||
| XM_027975772.1 | XP_027831573.1 | 24 - 25 | 3249 | 76 | ||
| XM_027975773.1 | XP_027831574.1 | 16 - 17 | 1896 | 76 | ||
| XM_027975774.1 | XP_027831575.1 | 16 - 17 | 1775 | 76 | ||
More Information
Set Membership: |
|

LOC101104843
Genomic Map