Assay Details
| Gene Symbol: SLC39A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967597.1 | XP_027823398.1 | 9 - 10 | 1479 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A5| Gene Symbol: SLC39A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967597.1 | XP_027823398.1 | 9 - 10 | 1479 | 60 | |
Set Membership: |
|