Assay Details
| Gene Symbol: KCNK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011627.4 | XP_004011676.2 | 1 - 2 | 309 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK9| Gene Symbol: KCNK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011627.4 | XP_004011676.2 | 1 - 2 | 309 | 128 | |
Set Membership: |
|