Assay Details
| Gene Symbol: LOC101104720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964663.1 | XP_027820464.1 | 9 - 10 | 2704 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104720| Gene Symbol: LOC101104720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964663.1 | XP_027820464.1 | 9 - 10 | 2704 | 73 | |
Set Membership: |
|