Assay Details
| Gene Symbol: LOC101104720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964663.1 | XP_027820464.1 | 4 - 5 | 1804 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104720| Gene Symbol: LOC101104720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964663.1 | XP_027820464.1 | 4 - 5 | 1804 | 90 | |
Set Membership: |
|