Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960091.1 | XP_027815892.1 | 4 - 5 | 553 | 85 | |
| XM_027960092.1 | XP_027815893.1 | 5 - 6 | 709 | 85 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map