Assay Details
| Gene Symbol: QRICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957660.1 | XP_027813461.1 | 5 - 6 | 1897 | 77 | |
| XM_027957661.1 | XP_027813462.1 | 4 - 5 | 1692 | 77 | ||
More Information
Set Membership: |
|

QRICH1
Genomic Map