Assay Details
| Gene Symbol: SHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969062.1 | XP_027824863.1 | 1 - 2 | 318 | 119 | |
| XM_027969063.1 | XP_027824864.1 | 1 - 2 | 318 | 119 | ||
More Information
Set Membership: |
|

SHH
Genomic Map