Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004563.4 | XP_004004612.2 | 4 | 2656 | 120 | |
| XM_027964818.1 | XP_027820619.1 | 4 | 2657 | 120 | ||
More Information
Set Membership: |
|

HOXD3
Genomic Map