Assay Details
| Gene Symbol: GPRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104306.2 | XP_014959792.2 | 3 | 1828 | 116 | |
| XM_027962185.1 | XP_027817986.1 | 2 | 2337 | 116 | ||
More Information
Set Membership: |
|

GPRIN2
Genomic Map