Assay Details
| Gene Symbol: LOC101111166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107009.2 | XP_011962399.2 | 1 | 2742 | 85 | |
| Gene Symbol: CBR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093286.2 | XP_014948772.2 | 5 | 2836 | 86 | |
More Information
Set Membership: |
|

LOC101111166
Genomic Map