Assay Details
| Gene Symbol: SRSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974149.1 | XP_027829950.1 | 2 | 757 | 83 | |
| XM_027974150.1 | XP_027829951.1 | 2 | 756 | 83 | ||
More Information
Set Membership: |
|

SRSF2
Genomic Map