Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008117.4 | XP_004008166.3 | 7 | 1265 | 110 | |
| XM_012177255.2 | XP_012032645.2 | 7 | 1386 | 110 | ||
More Information
Set Membership: |
|

SSBP1
Genomic Map