Assay Details
| Gene Symbol: PPP6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720442.3 | XP_002720488.1 | 487 | 94 | ||
| XM_017350096.1 | XP_017205585.1 | 810 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP6C| Gene Symbol: PPP6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720442.3 | XP_002720488.1 | 487 | 94 | ||
| XM_017350096.1 | XP_017205585.1 | 810 | 94 | |||
Set Membership: |
|