Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716486.3 | XP_002716532.1 | 7 - 8 | 874 | 64 | |
| XM_008266619.2 | XP_008264841.1 | 8 - 9 | 938 | 64 | ||
| XM_008266620.2 | XP_008264842.1 | 6 - 7 | 868 | 64 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map