Assay Details
| Gene Symbol: ENY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710737.3 | XP_002710783.1 | 2 - 3 | 226 | 97 | |
| XM_008255833.1 | XP_008254055.1 | 2 - 3 | 164 | 97 | ||
More Information
Set Membership: |
|

ENY2
Genomic Map