Assay Details
| Gene Symbol: NOL4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710921.2 | XP_002710967.1 | 1 - 2 | 502 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOL4L| Gene Symbol: NOL4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710921.2 | XP_002710967.1 | 1 - 2 | 502 | 77 | |
Set Membership: |
|