Assay Details
| Gene Symbol: CALU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082154.1 | NP_001075623.1 | 2 - 3 | 411 | 97 | |
| XM_008258168.2 | XP_008256390.1 | 3 - 4 | 528 | 97 | ||
| GenBank mRNA | AY225336.1 | - | 2 - 3 | 411 | 97 | |
More Information
Set Membership: |
|

CALU
Genomic Map