Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711756.3 | XP_002711802.1 | 13 - 14 | 1909 | 97 | |
| XM_008257735.2 | XP_008255957.1 | 13 - 14 | 1906 | 97 | ||
| GenBank mRNA | AB035256.1 | - | 13 - 14 | 1541 | 97 | |
More Information
Set Membership: |
|

GSPT1
Genomic Map