Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268556.2 | XP_008266778.1 | 2 | 978 | 80 | |
| XM_017347846.1 | XP_017203335.1 | 2 | 977 | 80 | ||
| GenBank mRNA | AF020291.1 | - | 1 | 351 | 80 | |
More Information
Set Membership: |
|

FMOD
Genomic Map