Assay Details
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195690.1 | NP_001182619.1 | 3 - 4 | 351 | 86 | |
| GenBank mRNA | AF069715.1 | - | 2 - 3 | 199 | 86 | |
More Information
Set Membership: |
|

TIMP4
Genomic Map