Assay Details
| Gene Symbol: LOC100008826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF413111.1 | - | 128 | 71 | ||
| EF583950.1 | - | 128 | 71 | |||
| Gene Symbol: LOC103347033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340383.1 | XP_017195872.1 | 409 | 71 | ||
More Information
Set Membership: |
|

LOC100008826
Genomic Map