Assay Details
| Gene Symbol: LOC100008826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF413111.1 | - | 271 | 82 | ||
| EF583950.1 | - | 271 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100008826| Gene Symbol: LOC100008826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF413111.1 | - | 271 | 82 | ||
| EF583950.1 | - | 271 | 82 | |||
Set Membership: |
|