Assay Details
| Gene Symbol: CXADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163251.1 | NP_001156723.1 | 4 - 5 | 567 | 152 | |
| GenBank mRNA | AY649414.1 | - | 3 - 4 | 429 | 152 | |
| EF034116.1 | - | 4 - 5 | 567 | 152 | ||
More Information
Set Membership: |
|

CXADR
Genomic Map