Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715991.3 | XP_002716037.1 | 4 - 5 | 441 | 78 | |
| XM_008265714.2 | XP_008263936.1 | 4 - 5 | 445 | 78 | ||
| GenBank mRNA | AY780571.1 | - | 3 - 4 | 289 | 78 | |
More Information
Set Membership: |
|

CDC42
Genomic Map