Assay Details
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716457.3 | XP_002716503.3 | 5 - 6 | 772 | 60 | |
| GenBank mRNA | D67014.1 | - | 5 - 6 | 667 | 60 | |
More Information
Set Membership: |
|

AHSG
Genomic Map