Assay Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195725.1 | NP_001182654.1 | 839 | 85 | ||
| GenBank mRNA | M26234.1 | - | 302 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195725.1 | NP_001182654.1 | 839 | 85 | ||
| GenBank mRNA | M26234.1 | - | 302 | 85 | ||
Set Membership: |
|