Assay Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337832.1 | XP_017193321.1 | 1124 | 79 | ||
| GenBank mRNA | EF420874.1 | - | 683 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A2| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337832.1 | XP_017193321.1 | 1124 | 79 | ||
| GenBank mRNA | EF420874.1 | - | 683 | 79 | ||
Set Membership: |
|