Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260511.2 | XP_008258733.1 | 4 | 818 | 85 | |
| GenBank mRNA | EU348828.1 | - | 1 | 353 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCR9| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260511.2 | XP_008258733.1 | 4 | 818 | 85 | |
| GenBank mRNA | EU348828.1 | - | 1 | 353 | 85 | |
Set Membership: |
|