Assay Details
| Gene Symbol: LPXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082048.1 | NP_001075517.1 | 7 - 8 | 818 | 66 | |
| GenBank mRNA | AF118146.1 | - | 7 - 8 | 818 | 66 | |
More Information
Set Membership: |
|

LPXN
Genomic Map