Assay Details
| Gene Symbol: MGST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082138.1 | NP_001075607.1 | 4 - 5 | 325 | 58 | |
| XM_008263951.2 | XP_008262173.1 | 6 - 7 | 481 | 58 | ||
| XM_008263952.2 | XP_008262174.1 | 5 - 6 | 436 | 58 | ||
| GenBank mRNA | AY050567.1 | - | 4 - 5 | 325 | 58 | |
More Information
Set Membership: |
|

MGST3
Genomic Map