Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082214.1 | NP_001075683.1 | 1 - 2 | 166 | 58 | |
| XM_008263962.2 | XP_008262184.1 | 1 - 2 | 222 | 58 | ||
| GenBank mRNA | DQ120011.1 | - | 1 - 2 | 166 | 58 | |
| GQ848313.1 | - | 1 - 2 | 166 | 58 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map