Assay Details
| Gene Symbol: MYLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082236.1 | NP_001075705.1 | 8 - 9 | 1678 | 82 | |
| GenBank mRNA | J05194.1 | - | 8 - 9 | 1678 | 82 | |
More Information
Set Membership: |
|

MYLK2
Genomic Map