Assay Details
| Gene Symbol: ALDOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082238.1 | NP_001075707.1 | 3 - 4 | 390 | 92 | |
| XM_008257929.2 | XP_008256151.1 | 3 - 4 | 479 | 92 | ||
| XM_008257930.2 | XP_008256152.1 | 3 - 4 | 461 | 92 | ||
| XM_017342433.1 | XP_017197922.1 | 3 - 4 | 506 | 92 | ||
| XM_017342435.1 | XP_017197924.1 | 3 - 4 | 484 | 92 | ||
| GenBank mRNA | K02300.1 | - | 3 - 4 | 390 | 92 | |
More Information
Set Membership: |
|

ALDOA
Genomic Map