Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082241.1 | NP_001075710.1 | 7 - 8 | 898 | 62 | |
| GenBank mRNA | L01791.1 | - | 7 - 8 | 898 | 62 | |
More Information
Set Membership: |
|

GYG1
Genomic Map