Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082259.1 | NP_001075728.1 | 2 - 3 | 1418 | 62 | |
| GenBank mRNA | L47207.1 | - | 2 - 3 | 1418 | 62 | |
More Information
Set Membership: |
|

PTGER4
Genomic Map