Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082293.1 | NP_001075762.1 | 2 - 3 | 231 | 114 | |
| GenBank mRNA | KT216053.1 | - | 2 - 3 | 202 | 114 | |
| M57439.1 | - | 2 - 3 | 231 | 114 | ||
More Information
Set Membership: |
|

CXCL8
Genomic Map