Assay Details
| Gene Symbol: PON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082297.1 | NP_001075766.1 | 6 - 7 | 745 | 83 | |
| GenBank mRNA | AF220941.1 | - | 5 - 6 | 619 | 83 | |
| AF220942.1 | - | 6 - 7 | 716 | 83 | ||
| AF220943.1 | - | 6 - 7 | 716 | 83 | ||
| M63011.1 | - | 6 - 7 | 745 | 83 | ||
| S64616.1 | - | 6 - 7 | 745 | 83 | ||
More Information
Set Membership: |
|

PON1
Genomic Map