Assay Details
| Gene Symbol: EEF1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082339.1 | NP_001075808.1 | 4 - 5 | 812 | 56 | |
| GenBank mRNA | U09823.1 | - | 4 - 5 | 812 | 56 | |
| X62245.1 | - | 782 | 56 | |||
More Information
Set Membership: |
|

EEF1A1
Genomic Map