Assay Details
| Gene Symbol: APOBEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082341.1 | NP_001075810.1 | 4 - 5 | 613 | 116 | |
| GenBank mRNA | U10695.1 | - | 4 - 5 | 613 | 116 | |
More Information
Set Membership: |
|

APOBEC1
Genomic Map