Assay Details
| Gene Symbol: ATP6V1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082368.1 | NP_001075837.1 | 6 - 7 | 572 | 82 | |
| GenBank mRNA | U45447.1 | - | 6 - 7 | 572 | 82 | |
More Information
Set Membership: |
|

ATP6V1D
Genomic Map