Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082640.1 | NP_001076109.1 | 2 - 3 | 209 | 66 | |
| GenBank mRNA | AY574214.1 | - | 2 - 3 | 209 | 66 | |
More Information
Set Membership: |
|

IMPACT
Genomic Map