Assay Details
| Gene Symbol: TGFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082660.1 | NP_001076129.1 | 6 - 7 | 1016 | 78 | |
| XM_008268050.2 | XP_008266272.1 | 5 - 6 | 2298 | 78 | ||
| GenBank mRNA | AY429466.1 | - | 6 - 7 | 1016 | 78 | |
More Information
Set Membership: |
|

TGFB2
Genomic Map