Assay Details
| Gene Symbol: CD80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082663.1 | NP_001076132.1 | 3 - 4 | 723 | 69 | |
| XM_008266820.2 | XP_008265042.1 | 4 - 5 | 69 | |||
| XM_008266822.2 | XP_008265044.1 | 4 - 5 | 69 | |||
| GenBank mRNA | D49843.1 | - | 3 - 4 | 723 | 69 | |
More Information
Set Membership: |
|

CD80
Genomic Map