Assay Details
| Gene Symbol: ADH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082624.1 | NP_001076093.1 | 3 - 4 | 355 | 92 | |
| GenBank mRNA | Y15406.1 | - | 355 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADH5| Gene Symbol: ADH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082624.1 | NP_001076093.1 | 3 - 4 | 355 | 92 | |
| GenBank mRNA | Y15406.1 | - | 355 | 92 | ||
Set Membership: |
|