Assay Details
| Gene Symbol: SLC5A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082699.1 | NP_001076168.1 | 488 | 69 | ||
| GenBank mRNA | U08813.1 | - | 488 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A10| Gene Symbol: SLC5A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082699.1 | NP_001076168.1 | 488 | 69 | ||
| GenBank mRNA | U08813.1 | - | 488 | 69 | ||
Set Membership: |
|